SML3736
Hypoxia probe 2
95% (UPLC)
Synonym(s):
5’-Methoxy-1’,2’-dimethyl-3-(1-((3-oxo-3H-phenoxazin-7-yl)oxy)ethyl)-1H-indole-4’,7’-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C25H20N2O6
Molecular Weight:
444.44
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
95% (UPLC)
form
powder
storage condition
protect from light
color
, Faint orange to red
solubility
DMSO: 2 mg/mL, clear (Warmed)
shipped in
wet ice
storage temp.
-10 to -25°C
Biochem/physiol Actions
Hypoxia probe 2 is a resorufin-based non-fluorescent hypoxia probe that undergoes cytochrome P450 reductase (PH51)-catalyzed reduction to yield the fluorescent reporter resorufin (ex 545 nm, em 582 nm) in conditions of 4% O2 and lower, allowing imaging hypoxia levels similar to those detected by pimonidazole without the use of antibodies. When employed together with Tokyo green-based hypoxia probe 4 (Tokyo green ex 480 nm, em 515 nm), the probe pair allows the simultaneous (two-color) imaging of different hypoxia levels in living cells.
Resorufin-based hypoxia probe that allows fluorescent imaging of hypoxia levels in live cells similar to those detected by pimonidazole without the use of antibodies.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Two Color Imaging of Different Hypoxia Levels in Cancer Cells.
Journal of the American Chemical Society, 145(4), 2572-2583 (2023)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service