Skip to Content
Merck
All Photos(1)

Key Documents

SML2843

Sigma-Aldrich

NS3623

≥98% (HPLC)

Synonym(s):

1-(3-Trifluoromethylphenyl)-3-[2-(1H-tetrazol-5-yl)-4-bromophenyl]urea, N-[4-Bromo-2-(2H-tetrazol-5-yl)phenyl]-N′-[3-(trifluoromethyl)phenyl]urea, NS 3623

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrF3N6O
CAS Number:
Molecular Weight:
427.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)NC(=O)Nc2c(cc(cc2)Br)c3[nH]nnn3

InChI

1S/C15H10BrF3N6O/c16-9-4-5-12(11(7-9)13-22-24-25-23-13)21-14(26)20-10-3-1-2-8(6-10)15(17,18)19/h1-7H,(H2,20,21,26)(H,22,23,24,25)

InChI key

JXPULDIATMTIIN-UHFFFAOYSA-N

Biochem/physiol Actions

NS3623 is a potent and selective activator of KV11.1 (hERG) and KV4.3 channels. It activates the IKr and Ito currents and exhibits antiarrhythmic activity. NS3623 may also act as a partial blocker of KV11.1 channels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biophysical characterization of the new human ether-a-go-go-related gene channel opener NS3623 [N-(4-bromo-2-(1H-tetrazol-5-yl)-phenyl)-N'-(3'-trifluoromethylphenyl)urea]
Hansen RS, Diness TG, Christ T, Wettwer E, Ravens U, Olesen SP, Grunnet M
Molecular Pharmacology, 70, 1319-1329 (2006)
Sergio Lainez et al.
Biochemical pharmacology, 150, 120-130 (2018-01-30)
Transient outward potassium current (Ito) contributes to early repolarization of many mammalian cardiac action potentials, including human, whilst the rapid delayed rectifier K+ current (IKr) contributes to later repolarization. Fast Ito channels can be produced from the Shal family KCNDE
Jacqueline A Treat et al.
Biochemical pharmacology, 169, 113608-113608 (2019-08-30)
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs) are used for many applications including safety pharmacology. However, a deficiency or complete absence of several K+ currents suggests repolarization reserve is low in hiPSC-CMs. We determined whether a dual Ito and IKr

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service