Skip to Content
Merck
All Photos(1)

Key Documents

SML0618

Sigma-Aldrich

NSC697923

≥98% (HPLC)

Synonym(s):

2-Nitro-5-tosylfuran, 2-[(4-Methylphenyl)sulfonyl]-5-nitro-Furan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO5S
CAS Number:
Molecular Weight:
267.26
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(c2[o]c(cc2)[N+](=O)[O-])c1ccc(cc1)C

InChI

1S/C11H9NO5S/c1-8-2-4-9(5-3-8)18(15,16)11-7-6-10(17-11)12(13)14/h2-7H,1H3

InChI key

GAUHIPWCDXOLCZ-UHFFFAOYSA-N

General description

NSC697923 stimulates apoptosis in p53 mutant neuroblastoma (NB) cells by triggering JNK (c-Jun N-terminal kinases) pathway. This ubiquitin-conjugating enzyme E2 N (UBE2N) inhibitor is considered as an important treatment option for neuroblastoma (NB) patients. NSC697923 prevents the multiplication and survival of diffuse large B-cell lymphoma (DLBCL) cells in vitro.[1]

Application

NSC697923 has been used as an inhibitor in STING-dependent NF-κB (nuclear factor-κB) activation after DNA damage, mediated by TRAF6 (TNF receptor-associated factor 6).[2]

Biochem/physiol Actions

NSC697923 is a cell-permeable and selective inhibitor of the ubiquitin-conjugating (E2) enzyme Ubc13-Uev1A that blocks the formation of the Ubc13 - ubiquitin thioester conjugate, and suppresses constitutive NF-kB activity in ABC-DLBCL cells. Also, NSC697923 inhibits the proliferation and survival of ABC-DLBCL and GCB-DLBCL cancer cells.
NSC697923 is a cell-permeable and selective inhibitor of the ubiquitin-conjugating (E2) enzyme Ubc13-Uev1A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Non-canonical Activation of the DNA Sensing Adaptor STING by ATM and IFI16 Mediates NF-κB Signaling after Nuclear DNA Damage.
Dunphy G, et al.
Molecular Cell, 71(5), 745-760 (2018)
J Cheng et al.
Cell death & disease, 5, e1079-e1079 (2014-02-22)
Neuroblastoma (NB) is the most common extracranial neoplasm in children. In NB, loss of p53 function is largely due to cytoplasmic sequestration rather than mutation. Ubiquitin-conjugating enzyme E2 N (UBE2N), also known as Ubc13, is an E2 ubiquitin-conjugating enzyme that
Curtis D Hodge et al.
ACS chemical biology, 10(7), 1718-1728 (2015-04-25)
Ubc13 is an E2 ubiquitin conjugating enzyme that functions in nuclear DNA damage signaling and cytoplasmic NF-κB signaling. Here, we present the structures of complexes of Ubc13 with two inhibitors, NSC697923 and BAY 11-7082, which inhibit DNA damage and NF-κB
Kim Ghilarducci et al.
The FEBS journal, 288(16), 4849-4868 (2021-03-03)
Protein ubiquitination has been historically associated with protein degradation, but recent studies have demonstrated other cellular functions associated with substrate ubiquitination. Among the RING-type ubiquitin E3 ligase enzymes present in the human genome, RNF167 is a transmembrane protein located in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service