OH30206084
Ohaus® Adventurer® Precision Balance
Model AX2201/E, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Recommended Products
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30206084
weighing capacity measuring range
2200 g, resolution: 0.1 g
weighing pan W × L
19.5 cm × 17.5 cm
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service