Skip to Content
Merck
All Photos(1)

Key Documents

N3534

Sigma-Aldrich

Anti-Neurabin I antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Neural-tissue Specific F-actin Binding Protein I, Anti-Protein Phosphatase 1 Regulatory Subunit 9A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 123 kDa

contains

0.05% sodium azide

species reactivity

rat

technique(s)

indirect immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Neurabin I is expressed in the central nervous system of rats.

Immunogen

recombinant coiled-coil domain of rat neurabin I.

Biochem/physiol Actions

Neurabin I has a role in the assembly of F-actin. It has been shown to be associated with stimulation of hepatocarcinogenesis in rats.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Kakehashi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 119(1), 61-72 (2010-10-12)
In the present study, protein lysates from microdissected glutathione S-transferase placental-form-positive (GST-P(+)) foci and hepatocellular carcinomas from livers of rats treated with N-diethylnitrosamine followed by phenobarbital at doses of 0 and 500 ppm in the diet for 10 and 33

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service