Key Documents
GE28-9889-37
Sigma-Aldrich
XK 16/20 Column
Synonym(s):
XK Column
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Pricing and availability is not currently available.
material
polypropylene column
packaging
1 ea of
General description
XK empty columns provide reliable and reproducible separations. The tube material is of borosilicate glass, which allows for visual inspection of the media bed. The columns are made of biocompatible materials that exhibit excellent chemical resistance. A broad range of column dimensions makes XK columns suitable for many different purification needs. Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations. An extensive range of spare parts is available, facilitating maintenance.
Features and Benefits
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page covers how and why to use Sepharose Big Beads for purification from crude, viscous samples.
This page covers using SOURCE™ medias for the purification of proteins, peptides, or oligonucleotides.
This page covers how and when to use Sepharose XL media for purification of proteins.
Here we discuss purification options when working with Sepharose® High Performance and we present some examples of media screening, capture and purification.
See All