Skip to Content
Merck
All Photos(1)

Key Documents

MOCHA

2111901, mouse skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

MOCHA, 02111901

biological source

mouse skin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Skin Fibroblast

Cell Line Description

Mocha is a naturally occuring mouse mutant with a null mutation in the delta subunit of AP-3 complex. In Mocha cells, the ß3 amd μ3 subunits coassmeble into a heterodimer whereas the sigma 3 subunit remains monomeric. The mouse has a similar phenotype to the human genetic disorder, Hermansky Pudlak Syndrome (HPS). In both mice and humans, AP-3 deficiency results in hypopigmentation because of abnormalities in melanosomes. The Mocha cell line can be used to investigate the assembly of AP-3 complexes and AP-3 function.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 5x1,000 to 1x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service