Skip to Content
Merck
All Photos(1)

Key Documents

PHR1820

Supelco

Triamterene Related Compound C

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,4-Diamino-7-hydroxy-6-phenylpteridine, 2,4-diamino-7-hydroxy-6-phenylpteridine, NSC 33416

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N6O
CAS Number:
Molecular Weight:
254.25
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1680030

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

triamterene

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[nH]1c2nc(nc(c2nc([c]1=O)c3ccccc3)N)N

InChI

1S/C12H10N6O/c13-9-8-10(18-12(14)16-9)17-11(19)7(15-8)6-4-2-1-3-5-6/h1-5H,(H5,13,14,16,17,18,19)

InChI key

USTXKGPQJJRCTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triamterene Related Compound C is a pharmaceutical impurity of a potassium-sparing diuretic, triamterene. Triamterene exhibits antihypertensive activity. It is used in combination with thiazide deuretic, such as benzthiazide for treating hypertension and edema associated with mild to moderate congestive heart failure.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Triamterene may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using reversed-phase high-performance liquid chromatography technique.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0841 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Validation of RP-HPLC Method for Simultaneous Estimation of Triamterene and Benzthiazide in Tablets
Chauhan CV, et al.
PharmaTutor, 2(6), 115-122 (2014)
High performance liquid chromatography for simultaneous determination of xipamide, triamterene and hydrochlorothiazide in bulk drug samples and dosage forms
El-Hay ASS, et al.
Acta pharmaceutica, 66(1), 109-118 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service