Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of block
manufacturer/tradename
NIST®
format
matrix material
General description
Other Notes
Certified for the analytes listed below.
Aluminum (Al), Magnesium (Mg), Cadmium (Cd), Manganese (Mn), Chromium (Cr), Nickel (Ni), Copper (Cu), Silicon (Si), Iron (Fe), Tin (Sn), Lead (Pb)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Cadmium (Cd ), Chromium (Cr), Copper (Cu), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Nickel (Ni), Silicon (Si), Tin (Sn)
See certificate for values and more details at nist.gov/SRM.
Aluminum (Al), Magnesium (Mg), Cadmium (Cd), Manganese (Mn), Chromium (Cr), Nickel (Ni), Copper (Cu), Silicon (Si), Iron (Fe), Tin (Sn), Lead (Pb)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Cadmium (Cd ), Chromium (Cr), Copper (Cu), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Nickel (Ni), Silicon (Si), Tin (Sn)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service