NIST3141A
Potassium (K) standard solution
NIST® SRM® 3141a
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
Related Categories
General description
SRM 3141A_cert
SRM 3141A_SDS
Application
Features and Benefits
- The certified value of the SRM was determined by calculating the mass fraction values of Potassium, which is obtained through two methods:
- Gravimetric preparation using Potassium Chloride
- Inductively coupled plasma optical emission spectrometry (ICP-OES).
- The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
- The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.
Other Notes
Potassium (K)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service