Skip to Content
Merck
All Photos(1)

Key Documents

BCR176R

Fly ash (trace elements)

BCR®, certified reference material

Synonym(s):

City waste incineration ash (trace elements)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Application

  • Trace Elements Solidification in Construction Materials: Research demonstrates the utility of wood fly ash combined with blast furnace slag for alkali-activation, effectively solidifying trace elements for use in composite materials, offering sustainable solutions for waste management in construction (Narani et al., 2024).
  • Arsenic Speciation in Agricultural Applications: The use of silicon-rich fly ash along with manganese or zinc oxides has been studied for its effect on arsenic speciation in paddy soils, highlighting its potential to mitigate arsenic toxicity under various soil management conditions (Wisawapipat et al., 2024).
  • Energy Efficient Utilization for Material Synthesis: Fly ash is utilized in the energy-efficient preparation of silicon-aluminum composite aerogel and gypsum, offering innovative methods for resource recovery from industrial waste (Li et al., 2023).

Analysis Note

For more information please see:
BCR176R

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Britta Vortmann-Westhoven et al.
Electrophoresis, 38(3-4), 540-546 (2016-11-22)
In this work, we present a novel electrophoretic method that was developed for the determination of lithium and transition metals in LiNi

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service