Skip to Content
Merck
All Photos(1)

Key Documents

BCR155

Dibenz[a,c]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13N
CAS Number:
Molecular Weight:
279.33
Beilstein:
18500
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2nc3c4ccccc4c5ccccc5c3cc2c1

InChI

1S/C21H13N/c1-6-12-20-14(7-1)13-19-17-10-3-2-8-15(17)16-9-4-5-11-18(16)21(19)22-20/h1-13H

InChI key

GUZBPGZOTDAWBO-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR155

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service