Skip to Content
Merck
All Photos(1)

Key Documents

BCR059A

Ti Al6 V4 alloy (O)

BCR®, certified reference material, disc

Synonym(s):

Titanium-aluminum-vanadium alloy, Oxygen in titanium-aluminium-vanadium alloy, Titanium-aluminum-vanadium alloy Ti with 6% Al and 4% V

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

form

disc

manufacturer/tradename

JRC

format

matrix material

SMILES string

[Al].[Ti].[V]

InChI

1S/Al.Ti.V

InChI key

GDAKJZDZIXHMSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified for the mass fractions of nitrogen and oxygen.

Analysis Note

For more information please see:
BCR059A

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service