Skip to Content
Merck
All Photos(1)

Key Documents

44795

Supelco

N,O-Bis(tert-butyldimethylsilyl)acetamide

for GC derivatization, LiChropur, ≥98.0% (GC)

Synonym(s):

N-[(1,1-Dimethylethyl)dimethylsilyl]ethanimidic acid (1,1-dimethylethyl)dimethylsilyl ester, BTBSA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H33NOSi2
CAS Number:
Molecular Weight:
287.59
Beilstein:
2096003
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

for GC derivatization

Quality Level

Assay

≥98.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

storage temp.

2-8°C

SMILES string

C/C(O[Si](C)(C)C(C)(C)C)=N\[Si](C)(C)C(C)(C)C

InChI

1S/C14H33NOSi2/c1-12(15-17(8,9)13(2,3)4)16-18(10,11)14(5,6)7/h1-11H3/b15-12+

InChI key

XFPQBAQQJIQJLY-NTCAYCPXSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Method for the t-Butyldimethylsilylation of Alcohols with N,O-Bis(t-butyldimethylsilyl)acetamide.
Johnson, D.A. and Taubner, L. M
Tetrahedron Letters, 37, 605-608 (1996)
T.P. Mawhinney et al.
The Journal of Organic Chemistry, 47, 3336-3336 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service