Skip to Content
Merck
All Photos(1)

Key Documents

1808

Sigma-Aldrich

Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3

1.4 % (w/v) in 95% ethanol

Synonym(s):

Methylene Blue solution, Ehrlich’s reagent III, Loeffler’s Methylene Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
Beilstein:
3641570
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

IVD

for in vitro diagnostic use

concentration

1.4 % (w/v) in 95% ethanol

storage temp.

15-25°C

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used as a nuclear counterstain in the various Lymphocyte Enzyme kits, Procedure No. 181

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Yan et al.
Cell research, 28(11), 1062-1076 (2018-10-10)
N6-methyladenosine (m6A) on mRNAs is critical for various biological processes, yet whether m6A regulates drug resistance remains unknown. Here we show that developing resistant phenotypes during tyrosine kinase inhibitor (TKI) therapy depends on m6A reduction resulting from FTO overexpression in
J How et al.
Gynecologic oncology, 137(3), 436-442 (2015-04-15)
With the debate over extent of lymphadenectomy in endometrial cancer, sentinel lymph node (SLN) mapping may provide a focused approach to evaluate the most relevant lymph nodes (LN) while minimizing the complications. We evaluated SLN mapping using filtered technetium(99), indocyanine
Swati Singh et al.
The Analyst, 139(14), 3600-3606 (2014-05-31)
The first gold-mercaptopropionic acid-polyethylenimine composite based electrochemical DNA biosensor was fabricated for the early detection of Streptococcus pyogenes infection in humans causing rheumatic heart disease (heart valve damage). No biosensor is available for the detection of rheumatic heart disease (RHD).
Inga Hoffmann et al.
Journal of lipid research, 55(10), 2113-2123 (2014-08-15)
The genome of the rice blast fungus Magnaporthe oryzae codes for two proteins with N-terminal dioxygenase (DOX) and C-terminal cytochrome P450 (CYP) domains, respectively. One of them, MGG_13239, was confirmed as 7,8-linoleate diol synthase by prokaryotic expression. The other recombinant
Bram Vivijs et al.
Applied and environmental microbiology, 80(19), 6054-6061 (2014-07-27)
Some members of the family Enterobacteriaceae ferment sugars via the mixed-acid fermentation pathway. This yields large amounts of acids, causing strong and sometimes even lethal acidification of the environment. Other family members employ the 2,3-butanediol fermentation pathway, which generates comparatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service