Skip to Content
Merck
All Photos(1)

Key Documents

07547

Supelco

6-Methoxy-2-naphthoic acid

pharmaceutical impurity standard

Synonym(s):

Naproxen impurity O (PhEur), 6-Methoxy-2-naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CO2H
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

API family

naproxen

Assay

≥96.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

201-206 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc2cc(ccc2c1)C(O)=O

InChI

1S/C12H10O3/c1-15-11-5-4-8-6-10(12(13)14)3-2-9(8)7-11/h2-7H,1H3,(H,13,14)

InChI key

YZBILXXOZFORFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used for preparation of arylamide inhibitors (HIV-1 integrase)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service