Skip to Content
Merck
All Photos(1)

Key Documents

10109339001

Roche

Sorbitol Dehydrogenase (SDH)

from sheep liver

Synonym(s):

SDH, Sorbitol Dehydrogenase, sorbitol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

biological source

sheep liver

form

lyophilized

specific activity

~40 units/mg protein (At 25 °C with D-fructose as the substrate.)

mol wt

115  kDa

packaging

pkg of 10 mg (60 mg lyophilizate)

manufacturer/tradename

Roche

concentration

0.16 mg/mL Biuret

technique(s)

activity assay: suitable

color

white

optimum pH

7.5

solubility

water: 60 mg/mL, clear, colorless

suitability

suitable for ELISA applications

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

ADH <0.00182%
GIDH <0.00152%
Glucose-DH <0.00182% (NAD as coenz.)
LDH <0.00167%
MDH <0.00198%
NADH oxidase 0.00532%

storage temp.

2-8°C

Gene Information

General description

L-iditol:NAD+ 5′-oxidoreductase
Sorbitol dehydrogenase (SDH) from sheep liver has a molar mass of 152kDa. The protein is a tetramer of four identical subunits. Each of these subunits has 355 amino acid residues, of which 10 are cysteine residues. Each subunit contains a zinc atom at the active site, which is associated with three protein ligands and a water molecule. The zinc atom associates with the oxygen of the sorbitol hydroxyl or of the fructose carbonyl interconverted during catalysis.

Application

Reduces L-iditol to L-sorbose. Also acts on D-glucitol and other closely related sugar alcohols. Allows the reduction of ketones to polyols (see aldolases for the synthesis of ketoses).

Biochem/physiol Actions

Sorbitol dehydrogenase (SDH) catalyzes the reversible NAD-linked conversion of D-sorbitol to D-fructose and forms a part of the sorbitol pathway, which is a crucial bypass to glycolysis for glucose metabolism. This pathway has been found to be associated with the accumulation of sorbitol in lens, which results in diabetic cataractogenesis. SDH also oxidizes various polyols and other secondary alcohols into their corresponding ketones.

Quality

Contaminants: <0.01% ADH, <0.02% GIDH and glucose dehydrogenase, each, <0.05% LDH and MDH, each.

Unit Definition

One unit (U) sorbitol dehydrogenase will reduce 1 mol of D-fructose in one minute at +25 °C and pH 7.6 [triethanolamine buffer; 150 mM fructose (nonsaturating concentration)]. The above assay consumes 1 mol of NADH per mol of D-sorbitol formed.

Physical form

Lyophilizate (60 mg contain 10 mg enzyme protein and 50 mg maltose).

Preparation Note

Activator: The reactions (oxidation or reduction) are fastest in Tris or triethanolamine buffer.
Stabilizers: Maltose is used as stabilizer.
Storage conditions (working solution): An aqueous solution is stable at 2 to 8 °C for several weeks.

Storage and Stability

Store at 2 to 8 °C. (Store the lyophilizate dry.)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R I Lindstad et al.
European journal of biochemistry, 210(2), 641-647 (1992-12-01)
The relations between the kinetic parameters for both sorbitol oxidation and fructose reduction by sheep liver sorbitol dehydrogenase show that a Theorell-Chance compulsory order mechanism operates from pH 7.4 to 9.9. This is supported by many parallels with the kinetics
R I Lindstad et al.
The Biochemical journal, 330 ( Pt 1), 479-487 (1998-04-16)
The substrate specificity of sheep liver sorbitol dehydrogenase has been studied by steady-state kinetics over the range pH 7-10. Sorbitol dehydrogenase stereo-selectively catalyses the reversible NAD-linked oxidation of various polyols and other secondary alcohols into their corresponding ketones. The kinetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service