Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB141A4

Sigma-Aldrich

Milli-Mark® Anti-SSEA3 Alexa 488 Antibody, clone MC-631

Milli-Mark®, from rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45
Pricing and availability is not currently available.

biological source

rat

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FUT4(2526)

Related Categories

General description

The cell-surface markers used routinely to identify undifferentiated human embryonic stem cells (hESCs) were initially characterized as markers for mESCs, mouse embryonic carcinomas (ECs), or human EC cells. Stage-specific embryonic antigen-3 (SSEA-3) and SSEA-4 are widely used as cell-surface markers to define both human and mouse ESCs. SSEA-3 and SSEA-4 are expressed on undifferentiated hESCs but not on undifferentiated mESCs.

Specificity

Reacts with the Stage-specific embryonic antigen-3 (SSEA-3) that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES).

Immunogen

4-8 cell stage mouse embryos

Application

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation
This Milli-Mark Anti-SSEA3 Alexa 488 Antibody, clone MC-631 is validated for use in FC for the detection of SSEA3 Alexa 488.

Quality

Evaluated by flow cytometry using 2102 Ep cells.

Physical form

Purified mouse monoclonal IgM conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
2102 Ep Cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Unconjugated antibody Cat. No. MAB4303-I is known to react with mouse.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service