Skip to Content
Merck
All Photos(1)

Key Documents

AB15568

Sigma-Aldrich

Anti-Aquaporin 7 Antibody

Chemicon®, from rabbit

Synonym(s):

AQP7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AQP7(364)

Specificity

Recognizes Aquaporin 7 (AQP7).

Immunogen

Purified peptide from the N-terminus of rat AQP7 (Accession number P56403).

Application

Anti-Aquaporin 7 Antibody detects level of Aquaporin 7 & has been published & validated for use in IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1:200-1:600 using ECL on rat and mouse kidney membrane lysate.
Immunohistochemistry on rat testis sections.
Immunocytochemistry: 1:800 using 3T3-L1 adipocytes.
Dilutions should be made using a carrier protein such as BSA (1-3%).
Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
ImmunoAffinity Purified
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongying Peng et al.
Fertility and sterility, 95(4), 1514-1517 (2010-12-07)
The aquaglyceroporin aquaporin 7 (AQP7) is dynamically expressed in mouse uteri undergoing decidualization after implantation. The expansion of AQP7 during uterine decidualization is associated with elevated uterine glycerol accumulation and glycerol kinase expression, suggesting that glycerol might be a potential
Verodia Charlestin et al.
Cancers, 16(15) (2024-08-10)
AQPs contribute to breast cancer progression and metastasis. We previously found that genetic inhibition of Aqp7 reduces primary tumor burden and metastasis in breast cancer. In this study, we utilized two AQP inhibitors, Auphen and Z433927330, to evaluate the efficacy
Rong Guo et al.
Nutrition & metabolism, 17, 69-69 (2020-08-22)
Obesity, characterized by the excessive accumulation of triglycerides in adipocytes and their decreased excretion from adipocytes, is closely related to various health problems. Ginsenoside Rb1 (Rb1), the most active component of the traditional Chinese medicine ginseng, has been reported to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service