Skip to Content
Merck
All Photos(1)

Key Documents

869039-M

Sigma-Aldrich

Prolactin, Human Pituitary, Iodination Grade

Synonym(s):

Prolactin, Human Pituitary, Iodination Grade, hPRL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
biological source:
human
form:
solid
Assay:
≥98% (weight)

biological source

human

Quality Level

description

Merck USA index - 14, 7779

Assay

≥98% (weight)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.5% hFSH (weight)
≤1% hGH, hLH, hTSH (weight)

color

white

pI 

5.73

solubility

acidified ethanol: soluble
methanol: soluble

storage temp.

−20°C

General description

Native, human prolactin from pituitary gland. Single chain peptide hormone required for lactation in mammals. Prolactin receptors are present in breast tissue, adrenals, ovary, testis, kidney, and liver. Potency: ≥30 IU/mg (WHO 3rd IS 84/500).

Biochem/physiol Actions

Potency: ≥30 IU/mg (WHO 3rd IS 84/500)≥ 30 IU/mg (WHO 1st IRP 75/504)

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Preparation Note

Prepared from tissues of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service