Skip to Content
Merck
All Photos(1)

Key Documents

8.21107

Sigma-Aldrich

Tetrapropyl orthotitanate

for synthesis

Synonym(s):

Tetrapropyl orthotitanate, Propyl titanate(IV), Titanic acid tetrapropyl ester, Titanium tetrapropylate, Orthotitanic acid tetrapropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2O)4Ti
CAS Number:
Molecular Weight:
284.22
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-411-3
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

bp

170 °C/36 hPa

transition temp

flash point 38 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Ti](OCCC)(OCCC)(OCCC)OCCC

InChI

1S/4C3H7O.Ti/c4*1-2-3-4;/h4*2-3H2,1H3;/q4*-1;+4

InChI key

HKJYVRJHDIPMQB-UHFFFAOYSA-N

Analysis Note

Assay (ex Ti): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.041 - 1.044
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service