Skip to Content
Merck
All Photos(1)

Key Documents

8.18017

Sigma-Aldrich

Cyclopropylamine

for synthesis

Synonym(s):

Cyclopropylamine, Aminocyclopropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N
CAS Number:
Molecular Weight:
57.09
MDL number:
UNSPSC Code:
12352116
EC Index Number:
212-142-2
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

275 °C

potency

445 mg/kg LD50, oral (Rat)

mp

-10 °C

transition temp

flash point 1 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NC1CC1

InChI

1S/C3H7N/c4-3-1-2-3/h3H,1-2,4H2

InChI key

HTJDQJBWANPRPF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.813 - 0.816
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service