Skip to Content
Merck
All Photos(1)

Key Documents

8.14990

Sigma-Aldrich

Allyl hexanoate

for synthesis

Synonym(s):

Allyl hexanoate, Allyl caproate, Hexanoic acid allyl ester, Caproic acid allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-642-4
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

218 mg/kg LD50, oral (Rat)
300 mg/kg LD50, skin (Rabbit)

transition temp

flash point 74 °C

density

0.887 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O(CC=C)C(=O)CCCCC

InChI

1S/C9H16O2/c1-3-5-6-7-9(10)11-8-4-2/h4H,2-3,5-8H2,1H3

InChI key

RCSBILYQLVXLJG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.888 - 0.892
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service