Skip to Content
Merck
All Photos(2)

Key Documents

8.08915

Sigma-Aldrich

Zirconium(IV) acetylacetonate

for synthesis

Synonym(s):

Zirconium(IV) acetylacetonate, Tetrakis(2,4-pentanedionato)zirconium(IV), Tetrakis(acetylacetonato)zirconium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O8Zr
CAS Number:
Molecular Weight:
487.66
MDL number:
UNSPSC Code:
12352103
EC Index Number:
241-510-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

powder

potency

719 mg/kg LD50, oral (Rat)

pH

7.2 (20 °C, 4.5 g/L in H2O)

mp

189-192 °C

solubility

14 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[Zr+4].O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C

InChI

1S/4C5H7O2.Zr/c4*1-4(6)3-5(2)7;/h4*3H,1-2H3;/q4*-1;+4

InChI key

DFPGFWYWSAYLCW-UHFFFAOYSA-N

Application

  • New metallo-organic complex of zirconium (IV): A precursor for nano structured ZrO2 (VG Prasanth et al., 2015).
  • Zirconium (IV) acetylacetonate as a ring-opening initiator: Mediating one-step synthesis of biodegradable polyacids (P Dobrzyński et al., 2019).
  • Decomposition insights using synchrotron radiation: Formation of volatile Zr-intermediates (S Grimm et al., 2022).
  • High-performance polymer solar cells: Employing zirconium acetylacetonate film as cathode buffer layer (Z Tan et al., 2014).
  • Preparation of zirconia nanofibers: Using electrospinning and calcination with zirconium acetylacetonate as precursor (VV Rodaev et al., 2019).

Analysis Note

Assay (ex Zr): ≥ 98.0 %
Melting range (lower value): ≥ 187 °C
Melting range (upper value): ≤ 192 °C
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service