Skip to Content
Merck
All Photos(1)

Key Documents

8.07059

Sigma-Aldrich

Diethyl oxalate

for synthesis

Synonym(s):

Diethyl oxalate, Oxalic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-464-1
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.27 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

410 °C

potency

400 mg/kg LD50, oral (Rat)

expl. lim.

0.42-2.67 % (v/v)

bp

182-186 °C

mp

-41 °C

transition temp

flash point 75 °C

density

1.08 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)C(=O)OCC

InChI

1S/C6H10O4/c1-3-9-5(7)6(8)10-4-2/h3-4H2,1-2H3

InChI key

WYACBZDAHNBPPB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.076 - 1.079
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service