Skip to Content
Merck
All Photos(1)

Key Documents

5.00506

Sigma-Aldrich

EPAC Inhibitor, ESI-09

Synonym(s):

EPAC Inhibitor, ESI-09, 3-(5- tert-butyl-isoxazol-3-yl)-2-((3-chlorophenyl)-hydrazono)-3-oxo-propionitrile, cAMP -GEF Inhibitor, Exchange Protein Directly Activated by cAMP Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15ClN4O2
CAS Number:
Molecular Weight:
330.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

Clc1cc(ccc1)N\N=C(\C(=O)c2n[o]c(c2)C(C)(C)C)/C#N

InChI

1S/C16H15ClN4O2/c1-16(2,3)14-8-12(21-23-14)15(22)13(9-18)20-19-11-6-4-5-10(17)7-11/h4-8,19H,1-3H3/b20-13+

InChI key

DXEATJQGQHDURZ-DEDYPNTBSA-N

General description

A cell-permeable compound that targets cAMP-binding domain of EPAC/cAMP-GEF and inhibits cAMP-dependent EPAC GEF activity (IC₅₀ = 1.4 and 3.2 µM, respectively, against EPAC2 and EPAC1).
A cell-permeable propionitrile compound that targets the CBD (cAMP-binding domain) of EPAC/cAMP-GEF with 4-fold higher affinity than cAMP and selectively inhibits cAMP-dependent EPAC GEF activity (IC50 = 1.4 and 3.2 M, respectively, against EPAC2 and EPAC1; [cAMP] = 25 M) with much reduced potency toward cAMP-dependent PKA activity (10% and 20% inhibition at 100 M, respectively, against 100 M cAMP-stimulated Type Iα and IIβ PKA activity). Shown to block both basal and 8-CPT-2′-O-Me-cAMP (Cat. No. 116833) prodrug 007-AM-, but not EGF-, stimulated Akt phosphorylations in serum-starved human AsPC-1 (≥185%, ≤85%, and <10% of basal thr308/Ser473 phosphorylation, respectively, with 0, 1, 10 M ESI-09; [007-AM] = 10 M) and EPAC-mediated insulin secretion from rat INS-1 cells (IC50<5 M; [007-AM] = 10 M).
A cell-permeable, propionitrile compound that targets the CBD (cAMP-binding domain) of EPAC/cAMP-GEF with 4-fold higher affinity than cAMP (IC50 = 10 vs. 39 µM, respectively, against 8-NBD-cAMP in competition EPAC2 binding assay) and selectively inhibits cAMP-dependent EPAC GEF activity (IC50 = 1.4 and 3.2 µM, respectively, against EPAC2 and EPAC1; [cAMP] = 25 µM) with much reduced potency toward cAMP-dependent PKA activity (10% and 20% inhibition at 100 µM, respectively, against 100 µM cAMP-stimulated Type Iα and IIβ PKA activity). Shown to block both basal and 8-CPT-2′-O-Me-cAMP (Cat. No. 116833) prodrug 007-AM-stimulated Akt thr308/Ser473 phosphorylations in serum-starved human pancreatic cancer AsPC-1 (%thr308/%Ser473 of basal phosphorylation = 200%/185%, 85%/65%, and <10%/<10%, respectively, with 0, 1, 10 µM ESI-09; [007-AM] = 10 µM) and rat pancreatic β-cell INS-1 without affecting EGF-stimulated Akt phosphorylations, resulting in effective inhibition of EPAC-mediated insulin secretion from INS-1 cells (by 57% and 100%, respectively, at 5 and 10 µM; [007-AM] = 10 µM) and pancreatic cancer cell invasion (~60% inhibition at 5 µM using AsPC-1 and PANC-1 cells).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Almahariq, M., et al. 2013. Mol Pharmacol.83, 122.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service