05-23-0310
Natriuretic Peptide, C-Type, Human and Porcine
Member of the natriuretic peptide family that shares structural homology with ANP and plays a role in the regulation of body fluid homeostasis, vascular tone, and vascular growth.
Synonym(s):
Natriuretic Peptide, C-Type, Human and Porcine, CNP, GLSKGCFGLKLDRIGSMSGLGC
About This Item
Recommended Products
Quality Level
Assay
≥96% (HPLC)
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
color
white to off-white
solubility
5% acetic acid: 1 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
S1SC[C@@H](C(=O)N[C@H](C(=O)NCC(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)NCC(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)NCC(=O)N[C@H](C(=O)NCC(=O)N[C@@H](C1)C(=O)O)CC(C)C)CO)CCSC)CO)[C@H](CC)C)CCCN\C(=N\[H])\N)CC(=O)O)CC(C)C)
InChI
1S/C93H157N27O28S3/c1-12-52(10)76-91(146)104-42-72(127)108-65(44-122)88(143)114-57(26-30-149-11)82(137)118-64(43-121)80(135)103-39-70(125)106-58(31-48(2)3)78(133)101-41-74(129)110-68(92(147)148)47-151-150-46-67(109-73(128)40-100-77(132)54(23-16-18-27-94)111-89(144)66(45-123)119-85(140)59(32-49(4)5)105-69(124)37-96)90(145)116-62(35-53-21-14-13-15-22-53)79(134)102-38-71(126)107-60(33-50(6)7)84(139)112-55(24-17-19-28-95)81(136)115-61(34-51(8)9)86(141)117-63(36-75(130)131)87(142)113-56(83(138)120-76)25-20-29-99-93(97)98/h13-15,21-22,48-52,54-68,76,121-123H,12,16-20,23-47,94-96H2,1-11H3,(H,100,132)(H,101,133)(H,102,134)(H,103,135)(H,104,146)(H,105,124)(H,106,125)(H,107,126)(H,108,127)(H,109,128)(H,110,129)(H,111,144)(H,112,139)(H,113,142)(H,114,143)(H,115,136)(H,116,145)(H,117,141)(H,118,137)(H,119,140)(H,120,138)(H,130,131)(H,147,148)(H4,97,98,99)/t52-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,76-/m0/s1
InChI key
UHSBKBYWHCBDCJ-DYGWFTAGSA-N
General description
Biochem/physiol Actions
Adenylate cyclase, particulate guanylate cyclase
Packaging
Warning
Sequence
Physical form
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service