Skip to Content
Merck
All Photos(1)

Key Documents

C-134

Supelco

Carisoprodol-13C3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C913C3H24N2O4
Molecular Weight:
263.31
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

This stable-labeled internal standard is suitable for carisoprodol testing or isotope dilution methods by LC/MS or GC/MS for applications in forensic analysis, urine drug testing, clinical toxicology, or pain prescription monitoring. Sold under trade names such as Soma and Sanoma, carisoprodol is a skeletal muscle relaxant that is often abused recreationally. Carisoprodol is purportedly used by drug users to potentiate the effects of other narcotics.

Application


  • Pharmacotherapy for Spine-Related Pain in Older Adults: This study explores various pharmacotherapeutic approaches, including Carisoprodol, for managing spine-related pain in older adults, emphasizing its efficacy and safety within geriatric care (Fu JL, Perloff MD, 2022).

  • Rapid Extraction and Qualitative Screening of 30 Drugs in Oral Fluid: Demonstrates the application of Carisoprodol in forensic toxicology by detailing methods for rapid drug screening in oral fluid, specifically for driving under the influence of drugs (DUID) cases (Coulter C, Garnier M, Moore C, 2022).

  • Single and Multiple Dose PK-PD Characterization for Carisoprodol: Discusses a clinical trial that characterizes the pharmacokinetics and pharmacodynamics of Carisoprodol in healthy volunteers, assessing the impact of the CYP2C19 phenotype on its metabolism (Calvo A, Alonso S, Prieto E, et al., 2022).

  • Tizanidine: Advances in Pharmacology & Therapeutics and Drug Formulations: Although focusing on Tizanidine, this review touches upon comparative analysis with other muscle relaxants like Carisoprodol, providing insights into therapeutic advancements and formulation strategies (Zhu LL, Wang YH, Zhou Q, 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service