Skip to Content
Merck
All Photos(1)

Key Documents

900405P

Avanti

pacFA GlcCer

Avanti Research - A Croda Brand 900405P, powder

Synonym(s):

D-glucosyl-β-1,1′ N-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H69N3O8
CAS Number:
Molecular Weight:
707.98
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (900405P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900405P

lipid type

click reagents

shipped in

dry ice

storage temp.

−70°C

SMILES string

C#CCCCC1(N=N1)CCCCCCCCC(N[C@](CO[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2)([H])[C@H](O)/C=C/CCCCCCCCCCCCC)=O

InChI

1S/C39H69N3O8/c1-3-5-7-8-9-10-11-12-13-14-15-18-21-25-32(44)31(30-49-38-37(48)36(47)35(46)33(29-43)50-38)40-34(45)26-22-19-16-17-20-24-28-39(41-42-39)27-23-6-4-2/h2,21,25,31-33,35-38,43-44,46-48H,3,5-20,22-24,26-30H2,1H3,(H,40,45)/b25-21+/t31-,32+,33+,35+

Biochem/physiol Actions

Photoactivatable and clickable fatty acid (pacFA) is useful to study the protein lipid interactions in living cells. pacFA serves as a parent molecule for the biosynthesis of bifunctional lipids.

Packaging

5 mL Amber Glass Screw Cap Vial (900405P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo profiling and visualization of cellular protein--lipid interactions using bifunctional fatty acids
Haberkant P, et al.
Angewandte Chemie (International Edition in English), 52(14), 4033-4038 (2013)
Mathias J Gerl et al.
PloS one, 11(4), e0153009-e0153009 (2016-04-23)
Cell membranes contain hundreds to thousands of individual lipid species that are of structural importance but also specifically interact with proteins. Due to their highly controlled synthesis and role in signaling events sphingolipids are an intensely studied class of lipids.
Per Haberkant et al.
Biochimica et biophysica acta, 1841(8), 1022-1030 (2014-01-21)
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as
In vivo profiling and visualization of cellular protein-lipid interactions using bifunctional fatty acids.
Per Haberkant et al.
Angewandte Chemie (International ed. in English), 52(14), 4033-4038 (2013-03-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service