Skip to Content
Merck
All Photos(1)

Key Documents

857500C

Avanti

17:0 cyclo acid

Avanti Research - A Croda Brand

Synonym(s):

Cis-9.,10-methylenehexadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H32O2
CAS Number:
Molecular Weight:
268.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (857500C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (857500C-1mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(CCCCCCCC1C(C1)CCCCCC)=O

InChI

1S/C17H32O2/c1-2-3-4-8-11-15-14-16(15)12-9-6-5-7-10-13-17(18)19/h15-16H,2-14H2,1H3,(H,18,19)

InChI key

MUZYOAHCGSIXJH-UHFFFAOYSA-N

General description

17:0 cyclo acid is a 17 carbon cyclopropane fatty acid found in E. coli.[1]

Application

17:0 cyclo acid has been used as a cyclopropane fatty acid constituent in the preparation of E. coli polar lipid extract.[2]

Biochem/physiol Actions

17:0 cyclo acid or cis-9,10-methylenehexadecanoic acid blocks the synthesis of phosphatidic acid by inhibiting the activity of the enzyme L-glycerol 3-phosphate acyltransferase from E. coli.[1]

Packaging

5 mL Clear Glass Sealed Ampule (857500C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of L-Glycerol 3-Phosphate Acyltransferase from Escherichia coil by cis-9, l0-Methylenehexadecanoic Acid
Kito, M, et al.
Test, 71(1), 99-105 (1972)
Physical properties of liposomes and proteoliposomes prepared from Escherichia coli polar lipids
White G F, et al.
Biochimica et Biophysica Acta - Biomembranes, 1468(1-2), 175-186 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service