Skip to Content
Merck
All Photos(2)

Key Documents

855779C

Avanti

22:0 Lyso PC

1-behenoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-docosanoyl-sn-glycero-3-phosphocholine; PC(22:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H62NO7P
CAS Number:
Molecular Weight:
579.79
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855779C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855779C

concentration

10 mg/mL (855779C-25mg)

shipped in

dry ice

storage temp.

−20°C

Application

22:0 Lyso PC has been used as a standard for liquid chromatography with tandem mass spectrometry (LC-MS/MS) analysis.

Packaging

5 mL Clear Glass Sealed Ampule (855779C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The peroxisomal AAA ATPase complex prevents pexophagy and development of peroxisome biogenesis disorders
Law KB, et al.
Autophagy, 13(5), 868-884 (2017)
Comprehensive untargeted lipidomic analysis using core-shell C30 particle column and high field orbitrap mass spectrometer
Narvaez-Rivas M and Zhang Q
Journal of Chromatography A, 1440, 123-134 (2016)
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal
Francesca Saitta et al.
Colloids and surfaces. B, Biointerfaces, 186, 110715-110715 (2019-12-17)
A fifteen-components model membrane that reflected the 80 % of phospholipids present in Insulin Secretory Granules was obtained and thermodynamic exploitation was performed, through micro-DSC, in order to assess the synergic contributions to the stability of a mixed complex system
Mónica Narváez-Rivas et al.
Journal of chromatography. A, 1440, 123-134 (2016-03-02)
The goal of untargeted lipidomics is to have high throughput, yet comprehensive and unambiguous identification and quantification of lipids. Novel stationary phases in LC separation and new mass spectrometric instruments capable of high mass resolving power and faster scanning rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service