Skip to Content
Merck
All Photos(2)

Key Documents

850854P

Avanti

16:0 Dimethyl PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N,N-dimethyl, powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N,N-dimethyl; PDME; 110742; phosphatidyldimethylethanolamine;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H78NO8P
CAS Number:
Molecular Weight:
720.01
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850854P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850854P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)[H])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H78NO8P/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-38(41)45-35-37(36-47-49(43,44)46-34-33-40(3)4)48-39(42)32-30-28-26-24-22-20-18-16-14-12-10-8-6-2/h37H,5-36H2,1-4H3,(H,43,44)

InChI key

SKWDCOTXHWCSGS-UHFFFAOYSA-N

Application

16:0 Dimethyl PE or phosphatidyl-N,N-dimethylethanolamine (PDME) has been used as a substrate for phosphatidylethanolamine (PE) N-methyl transferase.

Biochem/physiol Actions

16:0 Dimethyl PE or phosphatidyl-N,N-dimethylethanolamine (PDME) acts as an intermediate for the synthesis of phosphatidylcholine (PC).

Packaging

5 mL Amber Glass Screw Cap Vial (850854P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic mechanism of phosphatidylethanolamine N-methyltransferase.
Ridgway N, et al.
The Journal of Biological Chemistry, 263(32), 16864-16871 (1988)
Purification of phosphatidylethanolamine N-methyltransferase from rat liver.
Ridgway N, et al.
Test, 262(35), 17231-17239 (1987)
Rat brain phosphatidyl-N, N-dimethylethanolamine is rich in polyunsaturated fatty acids
Tacconi M, et al.
Journal of Neurochemistry, 45(3), 805-809 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service