Skip to Content
Merck
All Photos(2)

Key Documents

840513C

Avanti

18:1 Phosphatidylethanol

1,2-dioleoyl-sn-glycero-3-phosphoethanol (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H76O8PNa
CAS Number:
Molecular Weight:
751.00
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840513C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840513C

concentration

10 mg/mL (840513C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C41H77O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-40(42)46-37-39(38-48-50(44,45)47-6-3)49-41(43)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h19-22,39H,4-18,23-38H2,1-3H3,(H,44,45);/q;+1/p-1/b21-19-,22-20-;/t39-;/m1./s1

InChI key

AHVVHDCFEFPKMN-AOEFFCPUSA-M

General description

Phosphatidylethanol (PEth) is a phospholipid, that is generated in the presence of ethanol in cell membranes. PEths like PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 possesses a common phosphoethanol head. Two fatty acid chains of variable length and degree of saturation are bound to this head group.

Application

18:1 Phosphatidylethanol is suitable to use as a standard for the quantification of PEths in whole blood and in venous or capillary dried blood spots (DBS).

Biochem/physiol Actions

Phosphatidylethanols (PEths) serves as crucial biomarkers of alcohol consumption.

Packaging

5 mL Clear Glass Sealed Ampule (840513C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of phosphatidylethanol 16: 0/18: 1, 18: 1/18: 1, and 16: 0/16: 0 in venous blood and venous and capillary dried blood spots from patients in alcohol withdrawal and control volunteers
Kummer N, et al.
Analytical and Bioanalytical Chemistry, 408(3), 825-838 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service