Skip to Content
Merck
All Photos(1)

Key Documents

810106C

Avanti

NBD AA

Avanti Research - A Croda Brand 810106C

Synonym(s):

20-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino] arachidonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34 N4O5
CAS Number:
Molecular Weight:
482.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810106C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810106C

concentration

1 mg/mL (810106C-1mg)

lipid type

fluorescent lipids
bioactive lipids

shipped in

dry ice

storage temp.

−70°C

General description

The endocannabinoid pathway influences behavior, lipogenesis, embryologic development, and pathophysiological effects including pain, inflammation, stress, obesity, and non-alcoholic fatty liver disease. Endogenous endocannabinoids including arachidonoylethanolamide (anandamide, AEA) and 2-arachidonoylglycerol (2-AG) are derived from arachidonic acid (AA). Fluorescent NBD-AA and NBD-AEA analogues are direct probes that can be used for visualizing the uptake, intracellular trafficking, and targeting of these molecules by living cells, as well as measuring the binding of endocannabinoids to cytosolic binding proteins. Importantly, direct binding studies show the affinities of the fluorescent analogues are in the same range as the respective unlabeled ligands. Fluorescent endocannabinoid probes are useful for mapping the impact of various treatments, inhibitors, etc on endocannabinoid uptake and distribution in living cells.

Packaging

5 mL Amber Glass Screw Cap Vial/Argon/Foil Bag 810106C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service