Key Documents
Z680869
Duran® stirred reactor caps and accessories
folding magnetic stirrer with shaft
Synonym(s):
1200391
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 1200391
fitting
for for GL 45 stirred reactor systems
General description
These components allow for extra media bottles to be connected or lets sterile pressure compensation be possible. The parts are autoclaveable and the drive is provided by a standard magnetic stirrer.
Features and Benefits
  • Manufactured from chemically resistant materials: PP, PTFE, PEEK, and stainless steel
  • Use with standard magnetic stirrers up to 500 rpm and allows a significantly improved mixing performance compared to magnetic stir bars
  • The adjustable stirrer shaft can be moved to the optimal position in either 500 or 1000 ml DURAN® GL 45 bottles
  • Flexible: Use with the DURAN® GL 45 / GL 14 tubing connection components to connect 1.6, 3.0, 3.2, or 6.0 mm outer diameter semi-rigid tubing for the addition or removal of liquids or gas
  • Autoclavable
  • Not suitable for use with overhead lab stirrers
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)