Key Documents
Z680486
Duran® GL 45 connection system caps and accessories
insert for hose connection screw cap GL 14, 3.2 mm
Synonym(s):
1129817
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
white PTFE/silicone
manufacturer/tradename
DURAN® 1129817
parameter
-50-230 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)