Key Documents
Z558095
Spinner turbines for Bruker spectrometers
variable temperature, PET, diam. 5 mm
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Wilmad B-PEEK-5
diam.
5 mm
General description
Fiberglass turbines are made from high density GFK fiberglass and incorporate a single O-ring imbedded at the turbine top to hold the sample tube firmly into place. The PET spinner turbine for Bruker spectrometers incorporates two O-rings for ease of use and snug fit on the sample tube. The Bruker PET spinner turbine is recommended for 500 and 600 MHz NMR spectrometers.
VT operation of -150°C to +200°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)