Key Documents
Z406260
Sigma-Aldrich
Oxford® Sure/Seal storage valve-cap
pkg of 1 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
packaging
pkg of 1 ea
General description
For use with Aldrich products packed in Sure/Seal bottles. Screws over Sure/Seal crown cap to permit repeated dispensing of product via syringe while ensuring positive valved closure. Technical Information Bulletin AL-195, includes instructions for use of the valve.
Legal Information
Oxford is a registered trademark of Oxford Worldwide LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.