Key Documents
Z303356
Quickfit® double surface (Davies) condenser
L 150 mm, joint: ST/NS 19/26
Synonym(s):
C5/12/SC
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS19/26
manufacturer/tradename
Quickfit C 5/12/SC
availability
not available in USA and Canada
not available in USA and Canada
L
150 mm
surface area
170 cm2
General description
Increased surface area and effectiveness when compared with the corresponding Liebig condenser. Inner cooling surface gives a baffling effect, while outer cooling surface prevents creep and loss of vapour when used with low boiling liquids.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)