SLW3151/10
Pyrex® Volumetric flask, heavy duty, class A, USP/ISO/DIN tolerances
capacity 25 mL
Synonym(s):
Pyrex, SciLabware 3151/10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
Pyrex flask
Joint
joint ST/NS neck10/19
feature
class A USP/ISO/DIN tolerances
packaging
pkg of 5 ea
manufacturer/tradename
SciLabware 3151/10
Overall H
110 mm
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
General description
Class A, Pyrex
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards
- Each flask is laser etched with an individual serial number, batch code and date of manufacture
- Conformity ′H′, certified
- Manufactured from chemically resistant Pyrex glass
- Inscriptions in white enamel
- Polyethylene stopper
Features and Benefits
• Complies with ISO 1042 and DIN 12664• Manufactured from borosilicate glass 3.3• Calibrated to the tolerance required by the United States Pharmacopoeia (USP) which is tighter than those of ISO/DIN standards• Each flask is laser etched with an individual serial number batch code and date of manufacture• Certificate available to download online by entering the unique batch code, which can be found on the base of each flask• With Polyethylene stopper• Inscriptions in white enamel• 20mL and 5000mL sizes are supplementary to DIN range.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service