Key Documents
MT51340403
Mettler-Toledo Economic & Easy dissolved oxygen sensor
cable size 1.1 m
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
description
BNC connector
manufacturer/tradename
Mettler-Toledo 51340403
cable size
1.1 m
General description
Galvanic dissolved oxygen sensor without integrated temperature sensor, robust and easy to use.
Choose from one of the most recent versions:
Certificates of Analysis (COA)