MT51340298
Mettler Toledo InLab® 605 dissolved oxygen sensor
cable L 5 m
All Photos(1)
About This Item
UNSPSC Code:
41115600
NACRES:
NB.79
Recommended Products
manufacturer/tradename
Mettler-Toledo 51340298
cable L
5 m
General description
The InLab® 605 DO sensor incorporates the newest technology in polarographic dissolved oxygen determination. With the InLab® 605 dissolved oxygen sensor you will find the high accuracy and reproducibility that you have come to trust with METTLER TOLEDO products.
The InLab® 605 sensor can be used in combination with the SevenExcellence multichannel benchtop meter or the SevenGo 6 portable meters.
There are certain applications in which such sensors need to be rugged enough to withstand harsh conditions in factories, laboratories or out in the field. For this reason the membrane is protected by a steel mesh and the shaft is made out of glass fiber reinforced PPS (polyphenylene sulfide).
To go with the InLab® 605 METTLER TOLEDO offers the best of the best accessories: membrane kit, electrolyte, sealing kit and electrode weight.
The InLab® 605 sensor can be used in combination with the SevenExcellence multichannel benchtop meter or the SevenGo 6 portable meters.
There are certain applications in which such sensors need to be rugged enough to withstand harsh conditions in factories, laboratories or out in the field. For this reason the membrane is protected by a steel mesh and the shaft is made out of glass fiber reinforced PPS (polyphenylene sulfide).
To go with the InLab® 605 METTLER TOLEDO offers the best of the best accessories: membrane kit, electrolyte, sealing kit and electrode weight.
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.