GF65583311
Nickel-chromium alloy, Ni80Cr20
foil, 100 x 100mm, 0.005mm thickness, as rolled, not light tested (nlt)
Synonym(s):
NI050200
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
description
not light tested (nlt)
form
foil (as rolled)
manufacturer/tradename
Goodfellow 655-833-11
size × thickness
100 mm × 100 mm × 0.005 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service