GF19001562
Titanium-vanadium-chromium-aluminum alloy, Ti69V15Cr13Al3
foil, 50 x 50mm, 0.038mm thickness, annealed, light tight (lt)
Synonym(s):
TJ110350
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
Recommended Products
description
light tight (lt)
form
annealed (
quarter hard)
foil (
quarter hard)
manufacturer/tradename
Goodfellow 190-015-62
size × thickness
50 mm × 50 mm × 0.038 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service