Key Documents
A300C
Sigma-Aldrich
Printed Circuit Board Chip
A300C
About This Item
UNSPSC Code:
12352200
NACRES:
NC.13
material
FR-4 glass epoxy laminate substrate
copper ground plane
copper via(s)
metal pad(s) (bond)
metal pin
plastic header
feature
12 position(s) die
wirebondable with headers
manufacturer/tradename
A300C
Choose from one of the most recent versions:
Certificates of Analysis (COA)