Skip to Content
Merck
All Photos(1)

Key Documents

708925

Sigma-Aldrich

N-[[2-(Diphenylphosphino)phenyl]methylene]-3-methyl-L-valine sodium salt

95%

Synonym(s):

Na-tBu-DippAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25NNaO2P
CAS Number:
Molecular Weight:
425.44
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

powder

reaction suitability

reaction type: click chemistry

mp

199 °C (dec.)

SMILES string

[Na+].CC(C)(C)[C@H](\N=C\c1ccccc1P(c2ccccc2)c3ccccc3)C([O-])=O

InChI

1S/C25H26NO2P.Na/c1-25(2,3)23(24(27)28)26-18-19-12-10-11-17-22(19)29(20-13-6-4-7-14-20)21-15-8-5-9-16-21;/h4-18,23H,1-3H3,(H,27,28);/q;+1/p-1/b26-18+;/t23-;/m1./s1

InChI key

ZHRLPDTWOAIAKW-CCNYVGMNSA-M

Application

Used in asymmetric copper catalyzed conjugate addition[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wencel, J. et al.
Tetrahedron Asymmetry, 19, 1804-1804 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service