670308
(R)-2-(Methoxydiphenylmethyl)pyrrolidine
95% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H21NO
CAS Number:
Molecular Weight:
267.37
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95% (HPLC)
optical purity
ee: ≥99.5% (HPLC)
storage temp.
2-8°C
SMILES string
COC([C@H]1CCCN1)(c2ccccc2)c3ccccc3
InChI
1S/C18H21NO/c1-20-18(17-13-8-14-19-17,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17,19H,8,13-14H2,1H3/t17-/m1/s1
InChI key
CGUGCZSRPDCLBT-QGZVFWFLSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service