Skip to Content
Merck
All Photos(2)

Documents

64296

Sigma-Aldrich

(Trimethylsilyl)methanesulfonate

≥97%

Synonym(s):

Methanesulfonic acid trimethylsilyl ester, Trimethylsilyl mesylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CSO3OSi(CH3)3
CAS Number:
Molecular Weight:
168.29
Beilstein:
2076698
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.422 (lit.)
n20/D 1.422

bp

103-104 °C/25 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OS(C)(=O)=O

InChI

1S/C4H12O3SSi/c1-8(5,6)7-9(2,3)4/h1-4H3

InChI key

BCHASIUOFGDRIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Reduction of esters to ethers using boron difluoride triflate etherate
  • Elimination of C(8)-functional groups via regioselective dehydration
  • Dealkylsilylation of alumazene derivatives

Other Notes

Silylating agent

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J G Jun et al.
Carbohydrate research, 163(2), 247-261 (1987-06-15)
Several per-O-methylated D-glucans and D-fructans were used as models in an attempt to identify new catalysts for carrying out reductive cleavage. Included in these model studies were several D-glucans that contained 4-linked D-glucopyranosyl residues as well as one having a
H.H. Hergott et al.
Liebigs Ann. Chem., 1718-1718 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service