Skip to Content
Merck
All Photos(1)

Key Documents

407011

Sigma-Aldrich

2-(2-Methoxyethoxy)acetic acid

technical grade

Synonym(s):

O-(2-Methoxyethyl)glycolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH2CO2H
CAS Number:
Molecular Weight:
134.13
Beilstein:
1753567
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

245-250 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

COCCOCC(O)=O

InChI

1S/C5H10O4/c1-8-2-3-9-4-5(6)7/h2-4H2,1H3,(H,6,7)

InChI key

CLLLODNOQBVIMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service