Skip to Content
Merck
All Photos(1)

Documents

401528

Sigma-Aldrich

Bismuth(III) fluoride

≥99.99% trace metals basis

Synonym(s):

Trifluorobismuthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiF3
CAS Number:
Molecular Weight:
265.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

impurities

≤100.0 ppm Trace Metal Analysis

solubility

H2O: insoluble(lit.)

density

8.3 g/mL at 25 °C (lit.)

SMILES string

F[Bi](F)F

InChI

1S/Bi.3FH/h;3*1H/q+3;;;/p-3

InChI key

BRCWHGIUHLWZBK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service